The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009537.3 | NP_033563.2 | 2 - 3 | 942 | 99 |
GenBank mRNA | AK076149.1 | - | 3 - 4 | 555 | 99 |
AK166542.1 | - | 3 - 4 | 280 | 99 | |
BC055899.1 | - | 2 - 3 | 920 | 99 | |
M73963.1 | - | 2 - 3 | 942 | 99 | |
M74590.1 | - | 2 - 3 | 882 | 99 |