The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011719.4 | NP_035849.3 | 3 - 4 | 656 | 58 |
GenBank mRNA | AB073819.1 | - | 3 - 4 | 619 | 58 |
AF469004.1 | - | 3 - 4 | 604 | 58 | |
AK052708.1 | - | 3 - 4 | 657 | 58 | |
BC110482.1 | - | 3 - 4 | 648 | 58 |