The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243046.1 | NP_001229975.1 | 11 - 12 | 1134 | 111 |
NM_001243047.1 | NP_001229976.1 | 7 - 8 | 817 | 111 | |
NM_011807.3 | NP_035937.2 | 19 - 20 | 2293 | 111 | |
GenBank mRNA | AF388675.1 | - | 19 - 20 | 2072 | 111 |
AK030221.1 | - | 11 - 12 | 1136 | 111 | |
AK039754.1 | - | 7 - 8 | 682 | 111 |