The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013851.2 | NP_038879.2 | 1 - 2 | 209 | 60 |
GenBank mRNA | AF498362.1 | - | 1 - 2 | 209 | 60 |
AK138196.1 | - | 1 - 2 | 210 | 60 | |
AK173034.1 | - | 1 - 2 | 136 | 60 | |
BC089557.1 | - | 1 - 2 | 57 | 60 | |
BC141401.1 | - | 1 - 2 | 172 | 60 |