The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159563.1 | NP_001153035.1 | 7 - 8 | 767 | 92 |
NM_015824.4 | NP_056639.3 | 7 - 8 | 767 | 92 | |
GenBank mRNA | AB190257.2 | - | 7 - 8 | 734 | 92 |
AJ132830.1 | - | 7 - 8 | 749 | 92 | |
AK045388.1 | - | 7 - 8 | 742 | 92 | |
AK078115.1 | - | 2 - 3 | 282 | 92 | |
AK086579.1 | - | 7 - 8 | 770 | 92 | |
AK145324.1 | - | 7 - 8 | 746 | 92 | |
BC061252.1 | - | 7 - 8 | 738 | 92 |