The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023049.1 | NP_075536.1 | 9 - 10 | 2155 | 110 |
GenBank mRNA | AB221595.1 | - | 8 - 9 | 1765 | 110 |
AF155353.1 | - | 7 - 8 | 1622 | 110 | |
AK003566.1 | - | 2 - 3 | 355 | 110 | |
AK083400.1 | - | 7 - 8 | 1679 | 110 | |
AK155235.1 | - | 5 - 6 | 1869 | 110 | |
BC031161.1 | - | 9 - 10 | 2155 | 110 |