Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tmco1
Entrez Gene ID
68944
Gene Name
transmembrane and coiled-coil domains 1
Gene Aliases
1190006A08Rik, 4930403O06Rik, AA109065, AU022572, ESTM39
Chromosome Location
Chr.1: 167308670 - 167333978 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039483.1 NP_001034572.1 1 - 2 265 109
GenBank mRNA AK037709.1 - 1 - 2 200 109
AK162344.1 - 1 - 2 204 109
BC011457.1 - 1 - 2 139 109
BC019761.1 - 1 - 2 183 109
BC020098.1 - 1 - 2 195 109

View Full Product Details