The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_134151.4 | NP_598912.4 | 11 - 12 | 1557 | 67 |
GenBank mRNA | AK028785.1 | - | 11 - 12 | 1419 | 67 |
AK043837.1 | - | 11 - 12 | 1481 | 67 | |
AK076634.1 | - | 11 - 12 | 1402 | 67 | |
AK139579.1 | - | 11 - 12 | 1402 | 67 | |
AK145356.1 | - | 11 - 12 | 1450 | 67 | |
AK151880.1 | - | 11 - 12 | 1409 | 67 | |
AK169708.1 | - | 11 - 12 | 1333 | 67 | |
BC013552.1 | - | 11 - 12 | 1488 | 67 | |
BC022143.1 | - | 11 - 12 | 1452 | 67 | |
BC026615.1 | - | 11 - 12 | 1464 | 67 |