Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cd99l2
Entrez Gene ID
171486
Gene Name
CD99 antigen-like 2
Gene Aliases
AW548191, Mic2l1, Xap89
Chromosome Location
Chr.X: 71420060 - 71492869 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199349.1 NP_001186278.1 7 - 8 724 61
NM_138309.3 NP_612182.1 6 - 7 655 61
GenBank mRNA AK031264.1 - 7 - 8 481 61
AK080225.1 - 7 - 8 480 61
AK166275.1 - 7 - 8 475 61
AY078163.1 - 6 - 7 451 61
BC031736.1 - 6 - 7 430 61
EF516990.1 - 7 - 8 482 61

View Full Product Details