Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tmem40
Entrez Gene ID
94346
Gene Name
transmembrane protein 40
Gene Aliases
9030407H20Rik, AV091463, W64236
Chromosome Location
Chr.6: 115729131 - 115762466 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168256.1 NP_001161728.1 7 - 8 601 116
NM_001168257.1 NP_001161729.1 6 - 7 526 116
NM_001168258.1 NP_001161730.1 2 - 3 326 116
NM_001168259.1 NP_001161731.1 3 - 4 93 116
NM_144805.2 NP_659054.2 7 - 8 469 116
GenBank mRNA BC019416.1 - 7 - 8 431 116
BX511925.1 - 2 - 3 53 116
CT010303.1 - 6 - 7 399 116
CV561368.1 - 6 - 7 468 116
CX564997.1 - 7 - 8 500 116

View Full Product Details