The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144817.2 | NP_659066.1 | 1 - 2 | 110 | 64 |
GenBank mRNA | AF428262.1 | - | 1 - 2 | 58 | 64 |
BC021840.1 | - | 1 - 2 | 59 | 64 | |
BY730299.1 | - | 1 - 2 | 111 | 64 |