Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc22a7
Entrez Gene ID
108114
Gene Name
solute carrier family 22 (organic anion transporter), member 7
Gene Aliases
NLT, OAT2
Chromosome Location
Chr.17: 46432185 - 46438477 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144856.2 NP_659105.2 7 - 8 1364 67
GenBank mRNA AB069965.1 - 7 - 8 1276 67
AK082865.1 - 7 - 8 1365 67
AK143949.1 - 7 - 8 1352 67
BC013474.1 - 7 - 8 1341 67
BC024119.1 - 7 - 8 1328 67
BC025813.1 - 7 - 8 995 67
BC026597.1 - 7 - 8 1318 67
BC026598.1 - 7 - 8 1316 67

View Full Product Details