Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Oas2
Entrez Gene ID
246728
Gene Name
2'-5' oligoadenylate synthetase 2
Gene Aliases
Oasl11
Chromosome Location
Chr.5: 120730333 - 120749884 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145227.3 NP_660262.2 7 - 8 1658 59
GenBank mRNA AB067535.1 - 7 - 8 1663 59
AF418010.1 - 7 - 8 1598 59
BC043667.1 - 1 - 2 115 59
BC118538.1 - 7 - 8 1598 59

View Full Product Details