The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145835.2 | NP_665834.1 | 13 - 14 | 1856 | 59 |
GenBank mRNA | AF309072.1 | - | 13 - 14 | 1857 | 59 |
AK053515.1 | - | 12 - 13 | 1691 | 59 |