The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153128.2 | NP_694768.1 | 4 - 5 | 720 | 89 |
GenBank mRNA | AK034170.1 | - | 4 - 5 | 708 | 89 |
BC034514.1 | - | 4 - 5 | 720 | 89 |