Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ggnbp2
Entrez Gene ID
217039
Gene Name
gametogenetin binding protein 2
Gene Aliases
AI451896, C75939, D330017P12Rik, DIF-3, Zfp403
Chromosome Location
Chr.11: 84832361 - 84870738 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB064543.1 - 5 - 6 727 91
BC145605.1 - 4 - 5 635 91
BC150801.1 - 4 - 5 635 91

View Full Product Details