The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178357.3 | NP_848134.1 | 3 - 4 | 1377 | 65 |
GenBank mRNA | AJ459773.2 | - | 3 - 4 | 1223 | 65 |
AK036618.1 | - | 3 - 4 | 1363 | 65 | |
AK038397.1 | - | 3 - 4 | 1475 | 65 | |
AK050156.1 | - | 3 - 4 | 1379 | 65 | |
AK054170.1 | - | 3 - 4 | 1349 | 65 | |
AK082838.1 | - | 3 - 4 | 1366 | 65 | |
AK084913.1 | - | 3 - 4 | 1350 | 65 | |
AK085319.1 | - | 3 - 4 | 1352 | 65 | |
AK089044.1 | - | 3 - 4 | 1357 | 65 | |
AK161233.1 | - | 3 - 4 | 1369 | 65 | |
BC060642.1 | - | 3 - 4 | 1375 | 65 |