The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153501.2 | NP_705721.2 | 3 - 4 | 924 | 124 |
GenBank mRNA | AK082821.1 | - | 3 - 4 | 930 | 124 |
AK154080.1 | - | 3 - 4 | 888 | 124 | |
AK159175.1 | - | 3 - 4 | 916 | 124 | |
AK167861.1 | - | 3 - 4 | 632 | 124 | |
BC106184.1 | - | 3 - 4 | 878 | 124 |