Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Insig1
Entrez Gene ID
231070
Gene Name
insulin induced gene 1
Gene Aliases
1810013C12Rik, Insig-1
Chromosome Location
Chr.5: 28071412 - 28078662 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153526.5 NP_705746.1 2 - 3 512 64
GenBank mRNA AF527630.1 - 2 - 3 526 64
AK088380.1 - 2 - 3 559 64
AK135941.1 - 2 - 3 563 64
AK135989.1 - 2 - 3 565 64
AK139913.1 - 2 - 3 562 64
AK151186.1 - 2 - 3 562 64
BC079559.1 - 2 - 3 512 64
BC132167.1 - 2 - 3 520 64
BC132499.1 - 2 - 3 520 64

View Full Product Details