Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pygb
Entrez Gene ID
110078
Gene Name
brain glycogen phosphorylase
Gene Aliases
-
Chromosome Location
Chr.2: 150786796 - 150831748 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153781.1 NP_722476.1 2 - 3 419 71
GenBank mRNA AK032769.1 - 2 - 3 432 71
AK035673.1 - 2 - 3 148 71
AK134667.1 - 2 - 3 429 71
AK142716.1 - 2 - 3 428 71
AK147763.1 - 2 - 3 451 71
AK168381.1 - 2 - 3 429 71
AK169052.1 - 2 - 3 454 71
AK169631.1 - 2 - 3 433 71
BC031457.1 - 2 - 3 427 71
BC035283.1 - 2 - 3 419 71

View Full Product Details