Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Brca2
Entrez Gene ID
12190
Gene Name
breast cancer 2
Gene Aliases
Fancd1, RAB163
Chromosome Location
Chr.5: 150522021 - 150570147 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081001.2 NP_001074470.1 14 - 15 7698 61
NM_009765.3 NP_033895.2 14 - 15 7504 61
GenBank mRNA AK165522.1 - 4 - 5 833 61
BC059904.1 - 4 - 5 961 61
U82270.1 - 13 - 14 7190 61
U89652.1 - 13 - 14 7228 61

View Full Product Details