Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bpifa1
Entrez Gene ID
18843
Gene Name
BPI fold containing family A, member 1
Gene Aliases
LUNX, NASG, Plunc, SPLUNC1, SPURT
Chromosome Location
Chr.2: 154142880 - 154149217 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011126.3 NP_035256.2 3 - 4 454 74
GenBank mRNA BC054375.1 - 3 - 4 435 74
U69172.1 - 3 - 4 450 74

View Full Product Details