Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ddx3y
Entrez Gene ID
26900
Gene Name
DEAD (Asp-Glu-Ala-Asp) box polypeptide 3, Y-linked
Gene Aliases
8030469F12Rik, D1Pas1-rs1, Dby
Chromosome Location
Chr.Y: 1260715 - 1286628 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012008.2 NP_036138.1 5 - 6 524 101
GenBank mRNA AJ007376.1 - 5 - 6 491 101
AK154382.1 - 5 - 6 484 101
BC021453.1 - 5 - 6 489 101
BC051046.1 - 5 - 6 513 101

View Full Product Details