The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012008.2 | NP_036138.1 | 13 - 14 | 1586 | 126 |
GenBank mRNA | AJ007376.1 | - | 13 - 14 | 1553 | 126 |
AK154382.1 | - | 13 - 14 | 1546 | 126 | |
BC021453.1 | - | 13 - 14 | 1551 | 126 | |
BC051046.1 | - | 13 - 14 | 1575 | 126 |