The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164274.1 | NP_001157746.1 | 8 - 9 | 3262 | 90 |
NM_013720.2 | NP_038748.2 | 8 - 9 | 3262 | 90 | |
GenBank mRNA | AF205935.1 | - | 7 - 8 | 3076 | 90 |
AK032570.1 | - | 6 - 7 | 3006 | 90 | |
AK046252.1 | - | 8 - 9 | 3263 | 90 | |
AK139919.1 | - | 8 - 9 | 3215 | 90 | |
AK172962.1 | - | 8 - 9 | 3185 | 90 |