The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026014.3 | NP_080290.3 | 9 - 10 | 1553 | 65 |
GenBank mRNA | AB086655.1 | - | 9 - 10 | 1532 | 65 |
AF477481.1 | - | 9 - 10 | 1553 | 65 | |
AK012041.1 | - | 9 - 10 | 1530 | 65 | |
AK017609.1 | - | 9 - 10 | 1527 | 65 | |
AK028287.1 | - | 9 - 10 | 1525 | 65 | |
AK049163.1 | - | 9 - 10 | 1525 | 65 | |
AK076527.1 | - | 8 - 9 | 1635 | 65 | |
AK083015.1 | - | 9 - 10 | 1528 | 65 | |
AK200535.1 | - | 1 - 2 | 65 | 65 | |
BC013842.1 | - | 9 - 10 | 1508 | 65 | |
BC024485.1 | - | 6 - 7 | 804 | 65 | |
BC048076.1 | - | 9 - 10 | 1553 | 65 |