Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cpne3
Entrez Gene ID
70568
Gene Name
copine III
Gene Aliases
5430428M23Rik, 5730450C07Rik, C85108, CPN3
Chromosome Location
Chr.4: 19519252 - 19570104 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027769.2 NP_082045.1 1 - 2 102 57
GenBank mRNA AK017357.1 - 1 - 2 81 57
AK017651.1 - 1 - 2 107 57
AK036664.1 - 1 - 2 82 57
AK146324.1 - 1 - 2 79 57
AK220348.1 - 1 - 2 73 57
BB611055.1 - 1 - 2 100 57
BY717432.1 - 1 - 2 102 57

View Full Product Details