The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145857.2 | NP_665856.2 | 11 - 12 | 3043 | 78 |
GenBank mRNA | AF520774.1 | - | 11 - 12 | 3088 | 78 |
AK089843.1 | - | 11 - 12 | 3007 | 78 | |
AY160220.1 | - | 10 - 11 | 2972 | 78 | |
AY160221.1 | - | 11 - 12 | 3044 | 78 | |
BC044774.2 | - | 11 - 12 | 3070 | 78 |