Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam13a
Entrez Gene ID
58909
Gene Name
family with sequence similarity 13, member A
Gene Aliases
AI462012, D430015B01Rik, FAM13A1, Precm1
Chromosome Location
Chr.6: 58933536 - 59024502 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153574.2 NP_705802.1 15 - 16 1823 71
GenBank mRNA AJ510136.1 - 15 - 16 1809 71
AK036291.1 - 8 - 9 1030 71
AK052421.1 - 15 - 16 1764 71
BC005554.1 - 15 - 16 1728 71
BC054053.1 - 15 - 16 1797 71
BC060978.1 - 15 - 16 1760 71
BC080861.1 - 15 - 16 1823 71

View Full Product Details