The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_170671.2 | NP_733771.2 | 18 - 19 | 2753 | 71 |
GenBank mRNA | AK029929.1 | - | 17 - 18 | 2929 | 71 |
AK166780.1 | - | 11 - 12 | 1901 | 71 | |
BC027327.1 | - | 14 - 15 | 2114 | 71 | |
BC050808.1 | - | 7 - 8 | 1276 | 71 | |
BE979961.1 | - | 1 - 2 | 71 |