The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007924.2 | NP_031950.2 | 1 - 2 | 169 | 72 |
GenBank mRNA | AK159027.1 | - | 1 - 2 | 156 | 72 |
BC014816.1 | - | 1 - 2 | 139 | 72 | |
BC024894.1 | - | 1 - 2 | 138 | 72 | |
BY252808.1 | - | 1 - 2 | 153 | 72 | |
BY272058.1 | - | 1 - 2 | 170 | 72 | |
U80227.1 | - | 1 - 2 | 142 | 72 |