Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ell
Entrez Gene ID
13716
Gene Name
elongation factor RNA polymerase II
Gene Aliases
Ell1, Men
Chromosome Location
Chr.8: 70539675 - 70592858 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007924.2 NP_031950.2 6 - 7 896 68
GenBank mRNA AK159027.1 - 6 - 7 883 68
BC014816.1 - 6 - 7 866 68
BC024894.1 - 6 - 7 865 68
U80227.1 - 6 - 7 869 68

View Full Product Details