Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ets2
Entrez Gene ID
23872
Gene Name
E26 avian leukemia oncogene 2, 3' domain
Gene Aliases
AU022856, Ets-2
Chromosome Location
Chr.16: 95702407 - 95721049 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011809.3 NP_035939.3 7 - 8 1047 65
GenBank mRNA AK014602.2 - 7 - 8 969 65
AK030537.1 - 3 - 4 349 65
AK038348.1 - 7 - 8 1034 65
AK128933.1 - 8 - 9 921 65
AK143687.1 - 7 - 8 999 65
AK144974.1 - 7 - 8 998 65
AK145852.1 - 7 - 8 883 65
AK150175.1 - 7 - 8 976 65
AK152750.1 - 7 - 8 1001 65
AK153339.1 - 7 - 8 999 65
AK153478.1 - 7 - 8 999 65
BC005486.1 - 7 - 8 968 65
BC005504.1 - 7 - 8 908 65
CT010235.1 - 6 - 7 809 65
J04103.1 - 7 - 8 946 65

View Full Product Details