Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hgd
Entrez Gene ID
15233
Gene Name
homogentisate 1, 2-dioxygenase
Gene Aliases
Hgo, aku
Chromosome Location
Chr.16: 37580153 - 37632026 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013547.3 NP_038575.2 2 - 3 255 78
GenBank mRNA AK002584.1 - 1 - 2 127 78
AK050193.1 - 2 - 3 155 78
BC037628.1 - 2 - 3 169 78
BC055029.1 - 2 - 3 136 78

View Full Product Details