The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015766.2 | NP_056581.1 | 3 - 4 | 619 | 123 |
GenBank mRNA | AF013114.1 | - | 3 - 4 | 602 | 123 |
AK155023.1 | - | 3 - 4 | 582 | 123 | |
AK155903.1 | - | 3 - 4 | 474 | 123 | |
BC008209.1 | - | 3 - 4 | 567 | 123 |