The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023485.3 | NP_075974.3 | 1 - 2 | 173 | 146 |
GenBank mRNA | AJ251641.1 | - | 1 - 2 | 109 | 146 |
AK160756.1 | - | 1 - 2 | 163 | 146 |