Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc29a3
Entrez Gene ID
71279
Gene Name
solute carrier family 29 (nucleoside transporters), member 3
Gene Aliases
4933435C21Rik, AW987637, Ent3
Chromosome Location
Chr.10: 60712072 - 60752850 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023596.3 NP_076085.1 4 - 5 656 76
GenBank mRNA AF326986.1 - 4 - 5 667 76
AK034133.1 - 4 - 5 652 76
AK051317.1 - 4 - 5 681 76
BC137864.1 - 4 - 5 639 76
BC144983.1 - 4 - 5 639 76
CV557129.1 - 3 - 4 526 76

View Full Product Details