The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024478.2 | NP_077798.1 | 1 - 2 | 72 | 93 |
GenBank mRNA | AK048732.1 | - | 1 - 2 | 72 | 93 |
AK050445.1 | - | 1 - 2 | 67 | 93 | |
AK089113.1 | - | 1 - 2 | 68 | 93 | |
AK139561.1 | - | 1 - 2 | 260 | 93 | |
AK161688.1 | - | 1 - 2 | 85 | 93 | |
BC002284.1 | - | 1 - 2 | 122 | 93 |