Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sf3a1
Entrez Gene ID
67465
Gene Name
splicing factor 3a, subunit 1
Gene Aliases
1200014H24Rik, 5930416L09Rik, AI159724
Chromosome Location
Chr.11: 4160354 - 4182541 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026175.5 NP_080451.4 6 - 7 1014 81
GenBank mRNA AK004763.1 - 6 - 7 997 81
AK028829.1 - 6 - 7 966 81
AK029095.1 - 6 - 7 984 81
AK030223.1 - 6 - 7 1001 81
AK134178.1 - 6 - 7 993 81
AK160061.1 - 6 - 7 997 81
BC029753.1 - 6 - 7 978 81

View Full Product Details