Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Necap1
Entrez Gene ID
67602
Gene Name
NECAP endocytosis associated 1
Gene Aliases
1200016B17Rik, AI747569, mFLJ00061
Chromosome Location
Chr.6: 122874557 - 122888941 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026267.2 NP_080543.2 3 - 4 314 63
GenBank mRNA AK004805.1 - 3 - 4 315 63
AK005263.1 - 3 - 4 305 63
AK081598.1 - 3 - 4 315 63
AK168500.1 - 3 - 4 315 63
AK220236.1 - 3 - 4 300 63
BC119133.1 - 3 - 4 326 63
BC120841.1 - 3 - 4 397 63

View Full Product Details