Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pnrc2
Entrez Gene ID
52830
Gene Name
proline-rich nuclear receptor coactivator 2
Gene Aliases
0610011E17Rik, D4Bwg0593e
Chromosome Location
Chr.4: 135870917 - 135873846 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026383.3 NP_080659.1 2 - 3 376 78
GenBank mRNA AK011192.1 - 2 - 3 354 78
AK012073.1 - 2 - 3 360 78
AK013163.1 - 2 - 3 360 78
AK018384.1 - 2 - 3 324 78
AK075816.1 - 2 - 3 360 78
AK077403.1 - 2 - 3 241 78
AK208389.1 - 1 - 2 75 78
AK214826.1 - 1 - 2 75 78
AK219214.1 - 1 - 2 75 78
BC006598.1 - 2 - 3 349 78

View Full Product Details