The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159635.1 | NP_001153107.1 | 1 - 2 | 221 | 125 |
NM_026434.4 | NP_080710.1 | 1 - 2 | 221 | 125 | |
NR_027515.1 | 1 - 2 | 221 | 125 | ||
GenBank mRNA | AK008110.1 | - | 1 - 2 | 77 | 125 |
AK021243.1 | - | 1 - 2 | 102 | 125 | |
AK035348.1 | - | 1 - 2 | 129 | 125 | |
AK036252.1 | - | 1 - 2 | 102 | 125 | |
AK135109.1 | - | 1 - 2 | 129 | 125 | |
BC005540.1 | - | 1 - 2 | 115 | 125 | |
BY009724.1 | - | 1 - 2 | 128 | 125 | |
CB183820.1 | - | 1 - 2 | 292 | 125 |