Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ddx59
Entrez Gene ID
67997
Gene Name
DEAD (Asp-Glu-Ala-Asp) box polypeptide 59
Gene Aliases
1210002B07Rik, 4833411G06Rik
Chromosome Location
Chr.1: 136415271 - 136456190 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026500.3 NP_080776.1 2 - 3 972 71
GenBank mRNA AK004833.1 - 2 - 3 973 71
AK013179.1 - 2 - 3 934 71
AK133731.1 - 2 - 3 934 71
BC023840.1 - 2 - 3 899 71

View Full Product Details