The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026519.3 | NP_080795.1 | 4 - 5 | 637 | 63 |
GenBank mRNA | AK003668.1 | - | 4 - 5 | 605 | 63 |
AK012047.1 | - | 4 - 5 | 638 | 63 | |
AK078165.1 | - | 4 - 5 | 638 | 63 | |
AK167745.1 | - | 4 - 5 | 589 | 63 | |
BC051926.1 | - | 4 - 5 | 593 | 63 |