Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc14a1
Entrez Gene ID
108052
Gene Name
solute carrier family 14 (urea transporter), member 1
Gene Aliases
2610507K20Rik, 3021401A05Rik, UT-B, Utb1
Chromosome Location
Chr.18: 78100091 - 78142119 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028122.4 NP_082398.1 1 - 2 181 66
GenBank mRNA AJ420967.1 - 1 - 2 40 66
AK012066.1 - 1 - 2 31 66
BC058594.1 - 1 - 2 68 66
BC086673.1 - 1 - 2 77 66
BC100570.1 - 1 - 2 68 66
BY231473.1 - 1 - 2 181 66

View Full Product Details