Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc14a1
Entrez Gene ID
108052
Gene Name
solute carrier family 14 (urea transporter), member 1
Gene Aliases
2610507K20Rik, 3021401A05Rik, UT-B, Utb1
Chromosome Location
Chr.18: 78100091 - 78142119 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171010.1 NP_001164481.1 6 - 7 1063 89
NM_001171011.1 NP_001164482.1 5 - 6 798 89
NM_028122.4 NP_082398.1 5 - 6 858 89
GenBank mRNA AF448798.1 - 4 - 5 655 89
AJ420967.1 - 5 - 6 717 89
AK012066.1 - 5 - 6 708 89
AK041979.1 - 5 - 6 800 89
AK153891.1 - 6 - 7 968 89
BC058594.1 - 5 - 6 745 89
BC086673.1 - 5 - 6 754 89
BC100570.1 - 5 - 6 745 89

View Full Product Details