Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc14a1
Entrez Gene ID
108052
Gene Name
solute carrier family 14 (urea transporter), member 1
Gene Aliases
2610507K20Rik, 3021401A05Rik, UT-B, Utb1
Chromosome Location
Chr.18: 78100091 - 78142119 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171010.1 NP_001164481.1 8 - 9 1344 88
NM_001171011.1 NP_001164482.1 7 - 8 1079 88
NM_028122.4 NP_082398.1 7 - 8 1139 88
GenBank mRNA AF448798.1 - 6 - 7 936 88
AJ420967.1 - 7 - 8 998 88
AK012066.1 - 7 - 8 989 88
AK041979.1 - 7 - 8 1081 88
AK153891.1 - 8 - 9 1249 88
BC058594.1 - 7 - 8 1026 88
BC086673.1 - 7 - 8 1035 88
BC100570.1 - 7 - 8 1026 88

View Full Product Details