The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171010.1 | NP_001164481.1 | 9 - 10 | 1389 | 71 |
NM_001171011.1 | NP_001164482.1 | 8 - 9 | 1124 | 71 | |
NM_028122.4 | NP_082398.1 | 8 - 9 | 1184 | 71 | |
GenBank mRNA | AF448798.1 | - | 7 - 8 | 981 | 71 |
AJ420967.1 | - | 8 - 9 | 1043 | 71 | |
AK012066.1 | - | 8 - 9 | 1034 | 71 | |
AK041979.1 | - | 8 - 9 | 1126 | 71 | |
AK153891.1 | - | 9 - 10 | 1294 | 71 | |
BC058594.1 | - | 8 - 9 | 1071 | 71 | |
BC086673.1 | - | 8 - 9 | 1080 | 71 | |
BC100570.1 | - | 8 - 9 | 1071 | 71 |