The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028186.4 | NP_082462.3 | 3 - 4 | 363 | 87 |
GenBank mRNA | AF358136.1 | - | 3 - 4 | 313 | 87 |
AK008814.1 | - | 3 - 4 | 345 | 87 | |
AK081904.1 | - | 2 - 3 | 309 | 87 | |
AK145155.1 | - | 3 - 4 | 345 | 87 | |
AK146667.1 | - | 3 - 4 | 345 | 87 | |
AK158350.1 | - | 3 - 4 | 345 | 87 | |
AK165116.1 | - | 2 - 3 | 309 | 87 | |
BC019952.1 | - | 3 - 4 | 332 | 87 |