The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031388.2 | NP_113565.2 | 5 - 6 | 480 | 86 |
GenBank mRNA | AF285570.1 | - | 2 - 3 | 76 | 86 |
AK082829.1 | - | 5 - 6 | 86 | ||
AK133417.1 | - | 5 - 6 | 484 | 86 |